Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
Catalog No. B851925OR Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851925OR Orange
B851925BE Beige
B851925BL Blue
B851925GN Green
B851925GY Gray
B851925LA Lavender
B851925LG Light Green
B851925LI Lilac
B851925PK Pink
B851925TN Tan
B851925WH White
B851925YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. B851925OR Supplier Epredia™ Supplier No. B851925OR
Add to Cart
Add to Cart
This item is not returnable. View return policy
For biopsy tissue samples, these cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Pore Style Small Squares
Quantity 1000/Cs.
Closure Included Yes
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
Color Orange
Product Type TissueLoc Cassette IV with Color-Matched Lids