Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
Catalog No. B851954GN Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851954GN Green
B851954BE Beige
B851954BL Blue
B851954GY Gray
B851954LA Lavender
B851954LG Light Green
B851954LI Lilac
B851954OR Orange
B851954PK Pink
B851954TN Tan
B851954WH White
B851954YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. B851954GN Supplier Epredia™ Supplier No. B851954GN
Add to Cart
Add to Cart
For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Pore Style Slots
Closure Included Yes
Quantity 1000/Cs.
Packaging Type Loose Packed
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
Color Green
Product Type Slotted Cassette