Learn More
Ethos Biosciences TissueLock Paraffin 56 8kg
Catalog No. 5000819207
Encompass_Preferred
TissueLock 56 histologic paraffin wax is a tissue infiltration and embedding medium with added polymer Use in accordance with established tissue embedding and processing procedures Melting Point 56 58 C
Catalog No. 50-008-19207 Supplier Ethos Biosciences Supplier No. TL5608
TissueLock 56 histologic paraffin wax is a tissue infiltration and embedding medium with added polymer Use in accordance with established tissue embedding and processing procedures Melting Point 56 58 C