Learn More
StatLab Tissuetamp Embedding Tamper Set
SDP
Catalog No. 22021371
Change view
Click to view available options
Color:
Gray
Catalog No. Color
22-021-371 Gray
1 options
This item is not returnable. View return policy
Catalog No. 22-021-371 Supplier StatLab Supplier No. TTS2
Add to Cart
Add to Cart
This item is not returnable. View return policy
StatLab Tissuetamp Embedding Tamper Set
  • Designed in partnership with a histotech, the wide base of these proprietary tampers means reduced manual effort and risk for repetitive motion injury
  • Ergonomic, heavy-duty, tech-friendly, easy and efficient
  • The set comes with two ergonomic embedding tampers and an easy-to-use stand that will keep the tampers within reach
  • Is easily adaptable to any brand of embedding station hot plate
  • Includes 1 set and stand
Specifications
Product Type Tissue Tampers
Color Gray