Learn More
Andwin Scientific TISSUETEK VIP MEDIA/8X1KG/1CS
SDP
Catalog No. NC9605785
Encompass_Preferred
The Tissue-Tek V.I.P. permits superior sample preparation and less compression in sections as thin as 2. This product is ideal for processing and embedding. It does not leave plasticizer residue to clog paraffin lines and will not discolor or produce resin precipitate. It is sold in the quantity of 1 kilogram.
Catalog No. NC9605785 Supplier Andwin Scientific Supplier No. 4005
The Tissue-Tek V.I.P. permits superior sample preparation and less compression in sections as thin as 2. This product is ideal for processing and embedding. It does not leave plasticizer residue to clog paraffin lines and will not discolor or produce resin precipitate. It is sold in the quantity of 1 kilogram.