Learn More
Covaris Inc tissueTUBE TT2 & Plug Case (300)
SDP
Catalog No. NC2681893
Encompass
tissueTUBEs for sample collection, storage, pulverization, and transfer. For samples of mass 2 g. For use with the CP02 cryoPREP® Automated Dry Pulverizer and Holder tissueTUBE TT1 & tissueTUBE Extra Thick.
Catalog No. NC2681893 Supplier Covaris Inc Supplier No. 520034
Add to Cart
Add to Cart
Request a Quote
tissueTUBEs for sample collection, storage, pulverization, and transfer. For samples of mass 2 g. For use with the CP02 cryoPREP® Automated Dry Pulverizer and Holder tissueTUBE TT1 & tissueTUBE Extra Thick.