Learn More
MSC Miller Titan Roofing Fall Protection Kit, Universal, 50 Ft., 400 Lb. Capacity, 1 Ea.
Catalog No. 5000508795
Encompass
This roofing fall protection system kit provides essential equipment for safety when working at heights. It includes a full-body harness, a high-strength lifeline, a rope grab with a shock-absorbing lanyard, a roof anchor, and a waterproof storage container.
  • Includes a reusable stainless steel roof anchor bracket with a plated D-ring
  • Features a 16 mm (5/8 in.) diameter copolymer rope lifeline with a locking snap hook
  • Equipped with a rope grab featuring a permanently-attached 0.9 m (3 ft.) shock-absorbing lanyard
  • Designed for universal L/XL fit
  • Meets or exceeds OSHA and ANSI standard Z359.1-2007
  • Includes a waterproof storage container
Catalog No. 50-005-08795 Supplier MSC Supplier No. 66941485
This roofing fall protection system kit provides essential equipment for safety when working at heights. It includes a full-body harness, a high-strength lifeline, a rope grab with a shock-absorbing lanyard, a roof anchor, and a waterproof storage container.
  • Includes a reusable stainless steel roof anchor bracket with a plated D-ring
  • Features a 16 mm (5/8 in.) diameter copolymer rope lifeline with a locking snap hook
  • Equipped with a rope grab featuring a permanently-attached 0.9 m (3 ft.) shock-absorbing lanyard
  • Designed for universal L/XL fit
  • Meets or exceeds OSHA and ANSI standard Z359.1-2007
  • Includes a waterproof storage container