Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Miller Titan™ Shock-Absorbing Lanyard, 2 Legs, 6 ft., 400 lb. Capacity, 1 ea

Catalog No. 5000510018
Encompass

This two-leg shock-absorbing lanyard features a stretchable design that expands and contracts to minimize tripping hazards. Constructed from durable polyester webbing with a tubular polyester jacket, it provides reliable performance in fall arrest systems. This lanyard includes a built-in shock absorber to reduce peak arresting forces and is suitable for use with fall protection equipment.

  • Features a 6 ft. (1.8 m) standard length
  • Equipped with two legs for 100% tie-off capability
  • Includes a 3/4-in. (19 mm) locking snap hook for harness connection
  • Includes 2-1/2-in. (64 mm) locking rebar hooks for anchorage
  • Meets OSHA, ANSI, and CSA safety standards
  • Hardware meets 50-hour salt spray test requirements
  • Integrated visual load indicator

Catalog No. 50-005-10018 Supplier MSC Supplier No. 00877779
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This two-leg shock-absorbing lanyard features a stretchable design that expands and contracts to minimize tripping hazards. Constructed from durable polyester webbing with a tubular polyester jacket, it provides reliable performance in fall arrest systems. This lanyard includes a built-in shock absorber to reduce peak arresting forces and is suitable for use with fall protection equipment.

  • Features a 6 ft. (1.8 m) standard length
  • Equipped with two legs for 100% tie-off capability
  • Includes a 3/4-in. (19 mm) locking snap hook for harness connection
  • Includes 2-1/2-in. (64 mm) locking rebar hooks for anchorage
  • Meets OSHA, ANSI, and CSA safety standards
  • Hardware meets 50-hour salt spray test requirements
  • Integrated visual load indicator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.