Learn More
TA Instruments Titanium, 25 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580728 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Titanium Quick Change Plate (25 mm) is designed for Advanced Peltier Plates and ARES-G2 APS systems. Features corrosion resistance, durable construction, and secure placement for reproducible rheological measurements.
Catalog No. 50-005-80728 Supplier TA Instruments Supplier No. 402633.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Titanium Quick Change Plate (25 mm) is designed for Advanced Peltier Plates and ARES-G2 APS systems. Features corrosion resistance, durable construction, and secure placement for reproducible rheological measurements.