Learn More
Inorganic Ventures TITANIUM 30ML
SDP
Supplier:  Inorganic Ventures CGTI1030ML
Encompass_Preferred
Inorganic Ventures' 10,000 ppm (µg/mL) Titanium for ICP is a 125mL certified reference material set in a nitric acid /hydrofluoric acid matrix for stability. This titanium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC2827625