Learn More
TA Instruments Titanium, 40 mm Plate with Stainless Steel Shaft, Upper Plate for Use with Lower Peltier Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580638 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Titanium, 40 mm Plate with stainless steel shaft is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.
Catalog No. 50-005-80638 Supplier TA Instruments Supplier No. 520400.945
Waters TA Instruments HR X0, Titanium, 40 mm Plate with stainless steel shaft is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.