Learn More
Thermo Scientific™ Titanium Buckets for SureSpin™ 632 rotor
Catalog No. 75003037 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
17 mL
38.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75-003-037 38.5 mL
75-003-036 17 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-003-037 Supplier Thermo Scientific™ Supplier No. 75003037
Add to Cart
Add to Cart
Request a Quote
For use with SureSpin™ 632 rotor
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SureSpin 632
Capacity 38.5 mL
Material Titanium
Quantity 6/Pk.