Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium diisopropoxide bis(acetylacetonate)

Catalog No. 325252500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-525-2500ML 500 mL
32-525-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-525-2500ML Supplier Sigma Aldrich Supplier No. 325252500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium diisopropoxide bis (acetylacetonate) (TAA) may be used as a precursor to develop self assembled macroporous titania particles by evaporation. Commercial titanium(IV) bis(acetylacetonate) diisopropoxide was used to synthesize titanium acetylacetonate xerogels by drying, which could be further used to prepare titania films. TAA may be used as a hole blocking layer to fabricate solar cells.TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Specifications

CAS 17927-72-9
Density 0.995 g/mL (at 25°C)
Molecular Formula C16H28O6Ti
Refractive Index n20/D 1.4935
Linear Formula [(CH3)2 CHO]2 Ti(C5H7O2)2
Quantity 500 mL
MDL Number MFCD00000031
Synonym Diisopropoxytitanium bis(acetylacetonate) solution; TYZOR™ AA organic titanate; Ti(acac)2 OiPr2
Molecular Weight (g/mol) 364.26
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.