Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Titanium Dioxide
SCI_ED

Catalog No. s25818 Shop All Fisherbrand Products
Change view
Click to view available options
:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S25818 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25818
Only null left
Add to Cart
Add to Cart

Specifications

Color White
CAS 13463-67-7
Quantity 100 g
Assay Percent Range >98%
Molecular Formula O2Ti
Linear Formula TiO2
MDL Number MFCD00011269 MFCD00210650
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES O=[Ti]=O
IUPAC Name dioxotitanium
Molecular Weight (g/mol) 79.87
PubChem CID 26042
ChEBI CHEBI:32234
Formula Weight 79.88
Packaging Solid
Density 3.84 to 4.26g/mL
Chemical Name or Material Titanium Dioxide
Show More Show Less
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.