Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Titanium dioxide | 13463-67-7 | 99.0% | TiO2 | 1 KG
SDP

Catalog No. 5000210735
Encompass_Preferred

Titanium dioxide (TiO2) is an inorganic white powder used as a photosensitizer and photocatalyst. It is commonly used as a pharmaceutical excipient - for example, as a flow aid, coating agent, sunscreen ingredient, or colorant - and is also applied in environmental remediation such as water treatment, air purification, and self-cleaning materials. The product is supplied at 99.0% purity.

  • White inorganic powder with photocatalytic properties
  • Functions as a pharmaceutical excipient: flow aid, coating agent, sunscreen, colorant
  • Useful for environmental remediation: water treatment, air purification, self-cleaning surfaces
  • High purity (99.0%) suitable for research and formulation
  • Available as a 1 kg package

Catalog No. 50-002-10735 Supplier Medchemexpress LLC Supplier No. HYW0512711KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Titanium dioxide (TiO2) is an inorganic white powder used as a photosensitizer and photocatalyst. It is commonly used as a pharmaceutical excipient - for example, as a flow aid, coating agent, sunscreen ingredient, or colorant - and is also applied in environmental remediation such as water treatment, air purification, and self-cleaning materials. The product is supplied at 99.0% purity.

  • White inorganic powder with photocatalytic properties
  • Functions as a pharmaceutical excipient: flow aid, coating agent, sunscreen, colorant
  • Useful for environmental remediation: water treatment, air purification, self-cleaning surfaces
  • High purity (99.0%) suitable for research and formulation
  • Available as a 1 kg package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.