Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Dioxide, Technical, Spectrum™ Chemical
SDP

Catalog No. 18607420 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-420 500 g Poly Bottle
18-607-421 2.5 kg Poly Bottle
18-607-422 12 kg Poly Pail
18-607-423 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-420 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1081500GM
Add to Cart
Add to Cart

T1081, 13463-67-7, TiO2

Spectrum™ Chemical Titanium Dioxide, Technical , is the naturally available oxide of titanium also known as titania and is used as a pigment in paints, food coloring, cosmetics and in the production of pure titanium, electrical ceramics, catalysts and other chemical applications.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Poly Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.