Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Dioxide, Technical, Spectrum™ Chemical
SDP

Catalog No. 18607423 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-423 45 kg Poly Bottle
18-607-420 500 g Poly Bottle
18-607-421 2.5 kg Poly Bottle
18-607-422 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-423 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T108145KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T1081, 13463-67-7, TiO2

Spectrum™ Chemical Titanium Dioxide, Technical , is the naturally available oxide of titanium also known as titania and is used as a pigment in paints, food coloring, cosmetics and in the production of pure titanium, electrical ceramics, catalysts and other chemical applications.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 45 kg
Percent Purity 100%
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Titanium Dioxide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.