Learn More
Description
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Powder, Needles or Crystals |
| Quantity | 2.5 kg |
| Percent Purity | 99 to 100.5% |
| Grade | USP |
| Loss on Drying | 0.005 |
| Loss on Ignition | 0.005 |
| Packaging | Poly Bottle |
| Chemical Name or Material | Titanium Dioxide |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
