Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Dioxide, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607426 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-426 12 kg Poly Pail
18-607-427 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-426 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TI14012KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

TI140, 13463-67-7, TiO2

Spectrum™ Chemical Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.005
Loss on Ignition 0.005
Packaging Poly Pail
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.