Learn More
Analytical Sales & Services Titanium Electrode. No holes.
Catalog No. 5000805361
Encompass
Titanium Electrode (no holes)\nfor use as anode in Undivided Flow Cell\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each\n
\n
    \n
  • Strong; does not wear mechanically when assembling the cell
  • \n
  • Can be used as-is, as an electrode in the flow electrolysis cell
  • \n
  • Most common carrier used to deposit other materials
  • \n
  • Can be used as a carrier for DSA (usually Ruthenium(IV) Oxide [RuO₂])
  • \n
  • Easily plated
  • \n
  • Can be plated with a robust layer of Platinum, given the extremely high price of a solid Platinum electrode
  • \n
\n
Catalog No. 50-008-05361 Supplier Analytical Sales & Services Supplier No. F1088139
Titanium Electrode (no holes)\nfor use as anode in Undivided Flow Cell\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each\n
\n
    \n
  • Strong; does not wear mechanically when assembling the cell
  • \n
  • Can be used as-is, as an electrode in the flow electrolysis cell
  • \n
  • Most common carrier used to deposit other materials
  • \n
  • Can be used as a carrier for DSA (usually Ruthenium(IV) Oxide [RuO₂])
  • \n
  • Easily plated
  • \n
  • Can be plated with a robust layer of Platinum, given the extremely high price of a solid Platinum electrode
  • \n
\n