Learn More
Strem, An Ascensus Company CAS# 7440-32-6. 500 x 100mm. Titanium foil (99.6+%). MFCD00011264
SDP
Catalog No. 5090114469
Encompass
CAS# 7440-32-6. 500 x 100mm. Titanium foil (99.6+%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: 0.025 mm thick x 100 mm wide Strem# 93-2244. http://www.strem.com/catalog/v/93-2244/
Catalog No. 50-901-14469 Supplier Strem, An Ascensus Company Supplier No. 932244500MM
CAS# 7440-32-6. 500 x 100mm. Titanium foil (99.6+%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: 0.025 mm thick x 100 mm wide Strem# 93-2244. http://www.strem.com/catalog/v/93-2244/