Learn More
Fine Science Tools Semken Forceps, 15cm
SDP
Catalog No. NC9131310
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.1mm/ Alloy / Material: Stainless Steel/ Length: 15cm
Catalog No. NC9131310 Supplier Fine Science Tools Supplier No. 1100915
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.1mm/ Alloy / Material: Stainless Steel/ Length: 15cm