Learn More
Med Vet International Titanium Hemostatic Sterile Clips (Ligating Clips), Medium/Small, J0795B, 30 cart.of 6 clips
Supplier: Med Vet International JOR795B
Pure titanium annealed to a malleable consistency providing maximum vessel occlusion and zero memory to prevent tip opening after application. Biologically inert, radiolucent and non-magnetic for use with MRI or related scanning equipment. Chevron shape provides progressive and encompassing tip-to-tip closure before complete and secure occlusion of tissue or vessel. Diamond-shaped grooving and interlocking atraumatic teeth design of the clip ensures enhanced security in vessel closure. Striation is composed of small pyramidal heads, similar to tungsten inserts from needle-holders, insuring a firm, multidirectional, non-hurting vessel grip. Triangular-shaped wire fits perfectly into the custom grooves of our appliers to provide mechanical power during application of the clip. Specially designed surface treatment and annealing provide a secure atraumacity. The exclusive small/medium size has been designed to match specific intermediary vessel sizes requested by surgeons.