Titanium(II) Hydride, 99% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8918306

Quantity 5g

 View more versions of this product

Catalog No. AA8918306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7704-98-5
    Physical Form Powder
    Assay Percent Range 99%, metal basis
    Molecular Formula H2Ti
    Formula Weight 49.92
    MDL Number MFCD00151301
    Chemical Name or Material Titanium(II) Hydride

    Specifications

    Formula Weight 49.92
    Quantity 5g
    DOT Information DOT Class: 4.1
    EINECS Number 231-726-8
    Physical Form Powder
    Density 3.9
    Assay Percent Range 99%, metal basis
    Molecular Formula H2Ti
    MDL Number MFCD00151301
    Health Hazard 1 Flammable.
    Sensitivity Air, moisture sensitive
    CAS 7704-98-5
    Health Hazard 2 Take precautionary measures against static discharges.
    Chemical Name or Material Titanium(II) Hydride

    Take precautionary measures against static discharges.

    Flammable.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit