Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(II) hydride, Ti 94.0% min.

Catalog No. AA8918318
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8918318 50 g
AA8918306 5 g
AA8918330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8918318 Supplier Thermo Scientific Chemicals Supplier No. 08918318
Only null left
Add to Cart

CAS: 7704-98-5 | H2Ti

For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H{2}Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-98-5
Molecular Formula H2Ti
MDL Number MFCD00151301

Specifications

Melting Point 450°C (decomposition)
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%, metal basis
Solubility Information Soluble in water.
Formula Weight 49.92
Sensitivity Air and moisture sensitive
Density 3.9
Chemical Name or Material Titanium(II) Hydride
Hazard Category H228-H500
Hazard Statement Take precautionary measures against static discharges.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1; Packaging Group: II
EINECSNumber 231-726-8
RTECSNumber XR2130000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.