Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(III) sulfate, 20% in 1-4% sulfuric acid

Catalog No. AA3966430
Click to view available options
Quantity:
250 g
1 kg

Catalog No. AA3966430

Supplier: Thermo Scientific Chemicals 03966430

CAS: 10343-61-0 | O12S3Ti2 | 383.902 g/mol

Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10343-61-0
Molecular Formula O12S3Ti2
Molecular Weight (g/mol) 383.902
MDL Number MFCD00036317
InChI Key SOBXOQKKUVQETK-UHFFFAOYSA-H
Synonym titanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
PubChem CID 165866
IUPAC Name titanium(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]

Specifications

Physical Form Liquid
Quantity 250 g
UN Number UN3264
Merck Index 14,9476
Solubility Information Fully miscible with water.
Formula Weight 383.38
Concentration or Composition (by Analyte or Components) 20% in 1 to 4% sulfuric acid
Density 1.15 g/mL
Chemical Name or Material Titanium(III) sulfate
Hazard Category H315-H319-H350
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.