Learn More
Titanium(III) sulfate, 20% in 1-4% sulfuric acid
Description
Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 kg |
| UN Number | UN3264 |
| Merck Index | 14,9476 |
| Solubility Information | Fully miscible with water. |
| Formula Weight | 383.38 |
| Concentration or Composition (by Analyte or Components) | 20% in 1 to 4% sulfuric acid |
| Density | 1.15 g/mL |
| Chemical Name or Material | Titanium(III) sulfate |
Safety and Handling
| Hazard Category | H315-H319-H350 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.