Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) 2-ethylhexyloxide, VERTEC™ EHT, Thermo Scientific™

Catalog No. p-7059459
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44678AK 250 mL
AA44678K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44678AK Supplier Thermo Scientific Chemicals Supplier No. 44678AK

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Esterification catalyst for plasticizer production, transesterification catalyst for higher acrylates, and olefin polymerization catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-10-6
Molecular Formula C32H68O4Ti
Molecular Weight (g/mol) 564.76
MDL Number MFCD00015264,MFCD00015264
InChI Key KTXWGMUMDPYXNN-UHFFFAOYNA-N
Synonym tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt
PubChem CID 54603245
IUPAC Name titanium(4+) tetrakis(2-ethylhexan-1-olate)
SMILES [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]

Specifications

Boiling Point 194°C (0.25mmHg)
Melting Point <-25°C
Physical Form Liquid
Quantity 250 mL
Beilstein 3692023
Flash Point 71°C (160°F)
Formula Weight 564.79
Refractive Index 1.4820
Density 0.926g/mL
Chemical Name or Material Titanium (IV) 2-ethylhexyloxide, VERTEC™ EHT
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.