Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(IV) butoxide

Catalog No. 2441125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-411-25G 5 g
24-411-2500G 500 g
24-411-22KG 2 kg
24-411-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 24-411-25G Supplier Sigma Aldrich Supplier No. 2441125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to prepare nanosized titania in the anatase form and ferroelectric Bi4Ti3O12 thin films. Used in the preparation of nanocrystalline TiO2 powders at room temperature.TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Specifications

CAS 5593-70-4
Density 1.00 g/mL (at 20°C)
Boiling Point 206°C
Molecular Formula C16H36O4Ti
Linear Formula Ti(OCH2CH2CH2CH3)4
Refractive Index n20/D 1.491 (literature)
Quantity 5 g
MDL Number MFCD00009433
Synonym Orthotitanic acid tetrabutylester; TNBT; TYZOR™ TBT organic titanate; Tetrabutyl orthotitanate; Tetrabutyl titanate
Molecular Weight (g/mol) 340.32
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.