Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) Chloride (14% in Dichloromethane, ca. 1.0mol/L), TCI America™
SDP

Catalog No. T2052100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2052100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2052100ML Supplier TCI America Supplier No. T2052100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Titanium(IV) chloride solution at approximately 1.0 mol/L in dichloromethane for use as a laboratory reagent.

Chemical Identifiers

CAS 7550-45-0 , 75-09-2
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Physical Form Liquid
Quantity 100 mL
UN Number 3289
Formula Weight 189.67
Percent Purity 14.0% to 15.0% (w/w)
Grade Laboratory
Chemical Name or Material Titanium(IV) Chloride (14% in Dichloromethane, ca. 1.0mol/L)
EINECSNumber (1)-0262
RTECSNumber XR1925000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.