Learn More
Strem, An Ascensus Company CAS# 7550-45-0. 250g. Titanium(IV) chloride, 99%. MFCD00011267
SDP
Supplier:  Strem, An Ascensus Company 221150250G
Encompass
CAS# 7550-45-0. 250g. Titanium(IV) chloride, 99%. MFCD00011267. Molecular Weight: 189.73. Molecular Formula: TiCl4. Color/form: pale yellow liq. Strem# 22-1150. http://www.strem.com/catalog/v/22-1150/
Catalog No. 50-901-10775
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.