Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(Iv) Ethoxide

Catalog No. 8671050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-710-50ML 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 86-710-50ML Supplier Sigma Aldrich Supplier No. 8671050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(IV) ethoxide can be used:As an activator and dehydrating agent for by condensation of (S)-(+)-p-toluenesulfinamide with aldehydes and ketones to form sulfinimines.As a Ti4+ ion source to form complexes of titanium (IV) acrylate with methacrylic acid.As a catalyst to synthesize esters of sterically hindered alcohols via transesterification reactions.

Specifications

CAS 10442-39-4
Molecular Formula C19H23N7O2S
Linear Formula C19H23N7O2S
Refractive Index n20/D 1.505; n20/D 1.509
Quantity 25 mg
Synonym Tetraethyl orthotitanate; Tetraethyl titanate
Molecular Weight (g/mol) 413.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.