Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(IV) ethoxide

Catalog No. 24475950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-475-950G 50 g
24-475-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-475-950G Supplier Sigma Aldrich Supplier No. 24475950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium ethoxide is used widely as a precursor for the synthesis of Ti4+ ions titania nanoparticles by various techniques such as :in the sol-gel synthesis of free-standing ferroelectric lead zirconate titanate nanoparticlesspherical titania cores, produced either by hydrolysis of Ti(IV) ethoxide droplets in contact with water vapor or by preadsorption of the titanium alkoxides into/onto the clay mineral platelets and their subsequent hydrolysis and calcination.

Specifications

CAS 3087-36-3
Density 1.088 g/mL (at 25°C)
Boiling Point 150°C to 152°C
Molecular Formula C8H20O4Ti
Refractive Index n20/D 1.505 (literature)
Linear Formula Ti(OC2H5)4
Quantity 50 g
MDL Number MFCD00009071
Synonym Tetraethyl orthotitanate; Tetraethyl titanate
Molecular Weight (g/mol) 228.11
Biological Activity Central nervous system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.