Learn More
Strem, An Ascensus Company CAS# 3087-36-3. 5g. Titanium(IV) ethoxide (99.99%-Ti) PURATREM. MFCD00009071
SDP
Catalog No. 5090110783
Encompass
CAS# 3087-36-3. 5g. Titanium(IV) ethoxide (99.99%-Ti) PURATREM. MFCD00009071. Molecular Weight: 228.14. Molecular Formula: Ti(OC2H5)4. Color/form: xtl. to pale orange liq. Strem# 22-2209. http://www.strem.com/catalog/v/22-2209/
Catalog No. 50-901-10783 Supplier Strem, An Ascensus Company Supplier No. 2222095G
CAS# 3087-36-3. 5g. Titanium(IV) ethoxide (99.99%-Ti) PURATREM. MFCD00009071. Molecular Weight: 228.14. Molecular Formula: Ti(OC2H5)4. Color/form: xtl. to pale orange liq. Strem# 22-2209. http://www.strem.com/catalog/v/22-2209/