Learn More
Strem, An Ascensus Company CAS# 3087-36-3. 500g. Titanium(IV) ethoxide (contains 5-15% isopropanol). MFCD00009071
SDP
Catalog No. 5090114430
Encompass
CAS# 3087-36-3. 500g. Titanium(IV) ethoxide (contains 5-15% isopropanol). MFCD00009071. Molecular Weight: 228.14. Molecular Formula: Ti(OC2H5)4. Color/form: xtl. to pale yellow liq. Strem# 93-2209. http://www.strem.com/catalog/v/93-2209/
Catalog No. 50-901-14430 Supplier Strem, An Ascensus Company Supplier No. 932209500G
CAS# 3087-36-3. 500g. Titanium(IV) ethoxide (contains 5-15% isopropanol). MFCD00009071. Molecular Weight: 228.14. Molecular Formula: Ti(OC2H5)4. Color/form: xtl. to pale yellow liq. Strem# 93-2209. http://www.strem.com/catalog/v/93-2209/