Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) fluoride, 98%

Catalog No. p-4904559
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1154014 25 g
AA1154022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1154014 Supplier Thermo Scientific Chemicals Supplier No. 01154014
Only null left
Add to Cart

CAS: 7783-63-3 | TiF4

Titanium(IV) fluoride is used for preparation of glycosyl fluorides, glycosylation, preparation of fluorohydrins, addition of carbanions to aldehydes and imines. It is a catalyst for chemoselective synthesis and deprotection of geminal diacetates of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-63-3
Molecular Formula TiF4
MDL Number MFCD00042545

Specifications

Melting Point 284° (sublimation)
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98%
Solubility Information Insoluble in water.
Formula Weight 123.89
Odor Odorless
Sensitivity Moisture sensitive
Density 2.798
Chemical Name or Material Titanium(IV) fluoride
Show More Show Less
Hazard Category H302+H312+H332-H314
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-017-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.