Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(Iv) Isopropoxide

Catalog No. 2052732L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2 L
2.5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-527-32L 2 L
20-527-3100ML 100 mL
20-527-325L 2.5 L
20-527-31L 1 L
20-527-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 20-527-32L Supplier Sigma Aldrich Supplier No. 2052732L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(Iv) Isopropoxide

Specifications

CAS 546-68-9
Melting Point 14°C to 17°C
Density 0.96 g/mL at 20°C
Boiling Point 232°C
Molecular Formula C12H28O4Ti
Linear Formula Ti[OCH(CH3)2]4
Refractive Index n20/D 1.464
MDL Number MFCD00008871
Quantity 2 L
Synonym TTIP; Tetraisopropyl orthotitanate
Molecular Weight (g/mol) 284.22 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.