Alfa Aesar™ Titanium(IV) oxide, 20-35% in H2O colloidal dispersion

UV protection in clear coatings

Manufacturer: Alfa Aesar™ 4451736

Quantity 500g

 View more versions of this product

Catalog No. AA4451736

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13463-67-7
    Molecular Formula TiO2
    Formula Weight 79.9
    MDL Number MFCD00011269
    Chemical Name or Material Titanium (IV) oxide, Colloidal dispersion

    Specifications

    Formula Weight 79.9
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011269
    Boiling Point 368°C
    Quantity 500g
    Health Hazard 1 Warning
    Density 1.26g/mL
    TSCA TSCA
    Molecular Formula TiO2
    CAS 13463-67-7
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    Concentration 20 to 35% in H2O
    Chemical Name or Material Titanium (IV) oxide, Colloidal dispersion

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit