Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(IV) oxide, mixture of rutile and anatase

Catalog No. 70034725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-034-725G 25 g
70-034-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-034-725G Supplier Sigma Aldrich Supplier No. 70034725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(IV) oxide, mixture of rutile and anatase (TiO2) is a titania based nanoparticle solution that is dispersed in water.TiO2 suspensions can be used for a variety of applications such as:photo-catalytic applicationscosmetics, textile, and paintsfabrication of flexible humidity sensors

Specifications

Boiling Point 100°C
CAS 13463-67-7
Quantity 25 g
Molecular Formula O2Ti
Linear Formula TiO2
MDL Number MFCD00011269
Synonym Titanium dioxide
Molecular Weight (g/mol) 79.87
Percent Purity 99.5% trace metals basis
Density 1.3 g/mL ±0.2 (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.