Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(IV) oxide, rutile

Catalog No. 63726225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-726-225G 25 g
63-726-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-726-225G Supplier Sigma Aldrich Supplier No. 63726225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

May contain up to 5 wt. % Silicon dioxide as a surface coating.Contains small amount of anatase.

Specifications

CAS 1317-80-2
Quantity 25 g
Molecular Formula O2Ti
Linear Formula TiO2
MDL Number MFCD00011269
Synonym Titanium dioxide
Molecular Weight (g/mol) 79.87
Percent Purity 99.5% trace metals basis
Density 4.17 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.