Learn More
Strem, An Ascensus Company CAS# 13463-67-7. 25g. Titanium(IV) oxide, sintered lumps, 99.5%. MFCD00011269
SDP
Catalog No. 5090114426
Encompass
CAS# 13463-67-7. 25g. Titanium(IV) oxide, sintered lumps, 99.5%. MFCD00011269. Molecular Weight: 79.90. Molecular Formula: TiO2. Color/form: white lump Strem# 93-2208. http://www.strem.com/catalog/v/93-2208/
Catalog No. 50-901-14426 Supplier Strem, An Ascensus Company Supplier No. 93220825G
CAS# 13463-67-7. 25g. Titanium(IV) oxide, sintered lumps, 99.5%. MFCD00011269. Molecular Weight: 79.90. Molecular Formula: TiO2. Color/form: white lump Strem# 93-2208. http://www.strem.com/catalog/v/93-2208/