Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide sulfate sulfuric acid hydrate

Catalog No. p-4904560
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3959136 500 g
AA39591A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3959136 Supplier Thermo Scientific Chemicals Supplier No. 03959136
Only null left
Add to Cart

CAS: 123334-00-9 | TiOSO4.xH2O + H2SO4

Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-00-9
Molecular Formula TiOSO4.xH2O + H2SO4
MDL Number MFCD00011270
Synonym Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Chemical Name or Material Titanium(IV) oxide sulfate sulfuric acid hydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.