Titanium(IV) oxide sulfate sulfuric acid hydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 39591A3

Quantity 2kg

 View more versions of this product

Catalog No. AA39591A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13825-74-6
    Physical Form Powder
    Molecular Formula TiOSO4.xH2O + H2SO4
    MDL Number MFCD00011270
    Synonym Titanium(IV) oxysulfate-sulfuric acid

    Specifications

    Quantity 2kg
    Color White
    Physical Form Powder
    Molecular Formula TiOSO4.xH2O + H2SO4
    MDL Number MFCD00011270
    CAS 13825-74-6
    Synonym Titanium(IV) oxysulfate-sulfuric acid

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit