Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(Iv) Oxysulfate

Catalog No. 140236X1KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-023-6X1KG 6 x 1 kg
14-023-1KG 1 kg
14-023-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 14-023-6X1KG Supplier Sigma Aldrich Supplier No. 140236X1KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(IV) oxysulfate has been used as a precursor to synthesize a wide range of titanium dioxide (TiO2) photocatalysts. It can also be used in the synthesis of freeze dried titania foams, and WO3-loaded TiO2 photocatalyst applicable for the oxidation of trans-ferulic acid to vanillin.

Specifications

CAS 13825-74-6
Quantity 6 x 1 kg
Molecular Formula O5STi -+ xH2O
Linear Formula TiOSO4 -+ aq
MDL Number MFCD00149873
Synonym Titanyl sulfate
Molecular Weight (g/mol) 159.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.