Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(IV) oxysulfate solution

Catalog No. 495379250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-537-9250ML 250 mL
49-537-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-537-9250ML Supplier Sigma Aldrich Supplier No. 495379250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(IV) oxysulfate solution

Specifications

Boiling Point >100°C
CAS 13825-74-6
Quantity 250 mL
Molecular Formula O5STi
Linear Formula TiOSO4
MDL Number MFCD00216886
Synonym Titanium oxide sulfate; Titanium oxysulfate; Titanyl sulfate (TiO(SO4))
Molecular Weight (g/mol) 159.93
Percent Purity 99.99% trace metals basis
Density 1.361 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.