Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(Iv) (Triethanolaminato)Isopropoxide Solution

Catalog No. 388173100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-817-3100ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 38-817-3100ML Supplier Sigma Aldrich Supplier No. 388173100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Specifications

CAS 17933-03-8
Melting Point 160°C to 162°C
Molecular Formula C7H9BO2
Linear Formula CH3C6H4B(OH)2
Quantity 5 g
MDL Number MFCD00040198
Synonym TYZOR(R) TE organic titanate
Molecular Weight (g/mol) 135.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.