Learn More
Electron Microscopy Sciences EMS Titanium Alloy Mayo Scissors, Curved, 6-3/4"
SDP
Catalog No. 501931446
Encompass_Preferred
Mayo surgical scissors are made of standard grade titanium alloy. Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium Alloy instruments keep edges sharp longer than traditional instruments and are more stain resistant than stainless steel. Do not use bleach or other caustic chemicals.
Blade: Curved
OAL: 6-3/4"
This item is not returnable. View return policy
Catalog No. 50-193-1446 Supplier Electron Microscopy Sciences Supplier No. 7299638
Mayo surgical scissors are made of standard grade titanium alloy. Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium Alloy instruments keep edges sharp longer than traditional instruments and are more stain resistant than stainless steel. Do not use bleach or other caustic chemicals.
Blade: Curved
OAL: 6-3/4"