Learn More
Waters Corp Mixer Assembly, Titanium, 340 μL, 4.6 mm x 50 mm
Catalog No. 502392737 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement titanium mixer assembly has a volume of 340 μL and dimensions of 4.6 mm x 50 mm. It is specifically designed as a component for ACQUITY Arc Bio Systems.
  • Replacement mixer assembly
  • Volume: 340 μL
  • Dimensions: 4.6 mm x 50 mm
  • Compatible with ACQUITY Arc Bio Systems
  • Mechanical type
  • UPLC system type
  • Annual replacement schedule
  • One unit per package
Catalog No. 50-239-2737 Supplier Waters Corp Supplier No. 700011554
This replacement titanium mixer assembly has a volume of 340 μL and dimensions of 4.6 mm x 50 mm. It is specifically designed as a component for ACQUITY Arc Bio Systems.
  • Replacement mixer assembly
  • Volume: 340 μL
  • Dimensions: 4.6 mm x 50 mm
  • Compatible with ACQUITY Arc Bio Systems
  • Mechanical type
  • UPLC system type
  • Annual replacement schedule
  • One unit per package