Learn More
Waters Corp Mixer Assembly, Titanium, 680μL, 4.6 mm x 100 mm
Catalog No. 502391312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement mixer assembly is constructed from titanium and features a 680μL capacity with dimensions of 4.6 mm x 100 mm. It is specifically designed as a mechanical component for UPLC systems and is part of the ACQUITY Arc Bio Systems.
  • Constructed from titanium.
  • Features a 680μL capacity.
  • Measures 4.6 mm x 100 mm.
  • Designed for UPLC systems.
  • Part of ACQUITY Arc Bio Systems.
  • Recommended for annual replacement.
Catalog No. 50-239-1312 Supplier Waters Corp Supplier No. 700011553
This replacement mixer assembly is constructed from titanium and features a 680μL capacity with dimensions of 4.6 mm x 100 mm. It is specifically designed as a mechanical component for UPLC systems and is part of the ACQUITY Arc Bio Systems.
  • Constructed from titanium.
  • Features a 680μL capacity.
  • Measures 4.6 mm x 100 mm.
  • Designed for UPLC systems.
  • Part of ACQUITY Arc Bio Systems.
  • Recommended for annual replacement.