Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Nickel composite, -140+325 mesh, 99% (metals basis), Thermo Scientific™

Catalog No. AA4402722
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4402722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4402722 Supplier Thermo Scientific Chemicals Supplier No. 4402722

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium Nickel composite
Name Note Alloy Precursor
Avg. Mol. Wt. or Mol. Wt. Range Ti:Ni; 50:50 wt%
Quantity 100 g
Mesh Size −140+325 Mesh
Assay Percent Notes (metals basis)
MDL Number MFCD02091734
UN Number UN3089
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.